Gene Symbol: Pcdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013753.2 | NP_001013775.2 | 2, 3 | 5144 | ||
GenBank mRNA | AK133637.1 | - | 2, 3 | 200 | ||
AK142358.1 | - | 2, 3 | 5140 | |||
AK147265.1 | - | 2, 3 | 5146 | |||
AK158668.1 | - | 2, 3 | 2532 | |||
BC079850.1 | - | 2, 3 | 3123 | |||
BC089004.1 | - | 2, 3 | 3033 | 6841033 |