Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5SL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK001103.1 | - | 2 | 110 | ||
AK298197.1 | - | 2 | 142 | |||
AK301940.1 | - | 2 | 152 | |||
AK316313.1 | - | 2 | 116 | |||
BQ072604.1 | - | 2 | 132 | |||
DA057313.1 | - | 2 | 143 |