Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RADIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018059.4 | NP_060529.4 | 7 | 1804 | ||
GenBank mRNA | AB058752.1 | - | 7 | 1748 | ||
AK000997.1 | - | 3 | 242 | |||
AK001186.1 | - | 4 | 787 | |||
AK294745.1 | - | 6 | 1603 | |||
AK296325.1 | - | 6 | 1325 | |||
AK302549.1 | - | 1796 | ||||
AL136731.1 | - | 2 | 35 | |||
BC117317.1 | - | 6 | 1690 | 40125868 | ||
BC126311.1 | - | 6 | 1690 | 8992027 | ||
BC143526.1 | - | 6 | 1690 | 9052035 | ||
EF560723.1 | - | 6 | 1713 |