Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014099.1 | NP_001014121.1 | 3 | 1426 | ||
GenBank mRNA | BC079247.1 | - | 3 | 1426 | 7132885 | |
DQ088964.1 | - | 1 | 973 |
Set Membership: |
G-protein coupled receptor |