Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014116.1 | NP_001014138.1 | 9 | 1470 | ||
XM_006245998.2 | XP_006246060.1 | 8 | 1564 | |||
XM_006245999.2 | XP_006246061.1 | 9 | 1483 | |||
GenBank mRNA | BC079205.1 | - | 9 | 1470 | 7131778 | |
FQ215543.1 | - | 1 | 394 |