Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014125.1 | NP_001014147.1 | 15 | 1325 | ||
GenBank mRNA | BC088305.1 | - | 15 | 1325 | 7323988 |