Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014138.2 | NP_001014160.2 | 40 | 4119 | ||
XM_008770681.1 | XP_008768903.1 | 40 | 4955 | |||
XM_008770682.1 | XP_008768904.1 | 40 | 4952 | |||
XM_008770683.1 | XP_008768905.1 | 40 | 4949 | |||
XM_008770684.1 | XP_008768906.1 | 40 | 4931 | |||
XM_008770685.1 | XP_008768907.1 | 40 | 4925 | |||
XM_008770686.1 | XP_008768908.1 | 40 | 4919 | |||
XM_008770687.1 | XP_008768909.1 | 38 | 3989 | |||
GenBank mRNA | BC083918.1 | - | 16 | 2057 |