Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017378.1 | NP_001017378.1 | 10 | 1268 | ||
GenBank mRNA | BC083891.1 | - | 10 | 1268 | 7132551 |