Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017461.1 | NP_001017461.1 | 2 | 100 | ||
XM_006251446.2 | XP_006251508.1 | 2 | 216 | |||
XM_006251447.2 | XP_006251509.1 | 2 | 362 | |||
XM_006251448.2 | XP_006251510.1 | 2 | 151 | |||
GenBank mRNA | BC083811.1 | - | 2 | 100 | 7110182 | |
FQ222842.1 | - | 2 | 75 |