Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017461.1 | NP_001017461.1 | 11 | 1573 | ||
XM_006251446.2 | XP_006251508.1 | 11 | 1689 | |||
XM_006251447.2 | XP_006251509.1 | 11 | 1835 | |||
XM_006251448.2 | XP_006251510.1 | 11 | 1612 | |||
GenBank mRNA | BC083811.1 | - | 11 | 1573 | 7110182 | |
FQ222842.1 | - | 11 | 1548 |
Gene Symbol: LOC103693780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770414.1 | XP_008768636.1 | 475 |