Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024256.1 | NP_001019427.1 | 7 | 1208 | ||
GenBank mRNA | BC093402.1 | - | 7 | 1208 | 7389622 | |
FQ231796.1 | - | 5 | 894 |