Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759741.1 | XP_008757963.1 | 2, 3 | 2591 | ||
XM_008759742.1 | XP_008757964.1 | 3, 4 | 561 | |||
XM_008759743.1 | XP_008757965.1 | 2, 3 | 267 | |||
GenBank mRNA | BC097426.1 | - | 2, 3 | 253 | 7455141 |