Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198532.1 | NP_001185461.1 | 10 | 2030 | ||
NM_001198533.1 | NP_001185462.1 | 11 | 2256 | |||
NM_018002.3 | NP_060472.2 | 11 | 2256 | |||
NM_181354.4 | NP_851999.2 | 9 | 2086 | |||
XM_006716595.2 | XP_006716658.1 | 10 | 2025 | |||
XM_017013589.1 | XP_016869078.1 | 9 | 2055 | |||
XM_017013590.1 | XP_016869079.1 | 9 | 1896 | |||
XM_017013591.1 | XP_016869080.1 | 9 | 1986 | |||
XM_017013592.1 | XP_016869081.1 | 9 | 1733 | |||
XM_017013593.1 | XP_016869082.1 | 9 | 1960 | |||
XM_017013594.1 | XP_016869083.1 | 9 | 1960 | |||
XM_017013595.1 | XP_016869084.1 | 8 | 1870 | |||
XM_017013596.1 | XP_016869085.1 | 10 | 2043 | |||
XM_017013597.1 | XP_016869086.1 | 8 | 1704 | |||
XM_017013598.1 | XP_016869087.1 | 8 | 1775 | |||
XM_017013599.1 | XP_016869088.1 | 8 | 1870 | |||
XM_017013600.1 | XP_016869089.1 | 8 | 1727 | |||
GenBank mRNA | AF309387.1 | - | 3 | 473 | ||
AK000987.1 | - | 2 | 227 | |||
AK292899.1 | - | 9 | 2038 | |||
AK296561.1 | - | 10 | 2030 | |||
AL833193.1 | - | 8 | 1667 | |||
BC032710.2 | - | 9 | 2028 | 5496492 | ||
BC035484.1 | - | 9 | 2040 | |||
FN650108.1 | - | 10 | 2066 |