Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025126.1 | NP_001020297.1 | 9 | 1067 | ||
XM_006250113.2 | XP_006250175.1 | 16 | 2266 | |||
XM_006250114.2 | XP_006250176.1 | 16 | 2254 | |||
XM_006250115.2 | XP_006250177.1 | 15 | 2206 | |||
XM_006250116.2 | XP_006250178.1 | 15 | 2194 | |||
GenBank mRNA | BC091284.1 | - | 9 | 1067 | 7304516 |