Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP19 SIRNA Assays ›
Gene Symbol:
USP19
Gene Name:
ubiquitin specific peptidase 19
Gene Aliases:
ZMYND9
Chromosome Location:
Chr.3: 49108046 - 49120938 on Build GRCh38
UniGene ID:
Hs.255596
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
261015
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP19
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199160.1
NP_001186089.1
13, 14
2343
NM_001199161.1
NP_001186090.1
13, 14
2349
NM_001199162.1
NP_001186091.1
13, 14
2313
NM_006677.2
NP_006668.1
12, 13
2040
XM_005264823.2
XP_005264880.1
13, 14
2350
XM_005264824.2
XP_005264881.1
13, 14
2369
XM_005264825.2
XP_005264882.1
13, 14
2344
XM_005264826.2
XP_005264883.1
13, 14
2344
XM_005264827.2
XP_005264884.1
13, 14
2305
XM_005264829.2
XP_005264886.1
13, 14
2350
XM_005264830.2
XP_005264887.1
12, 13
2047
XM_005264831.2
XP_005264888.1
12, 13
1996
XM_006712946.2
XP_006713009.1
13, 14
2347
XM_006712947.2
XP_006713010.1
13, 14
2296
XM_006712948.2
XP_006713011.1
13, 14
2338
XM_006712949.2
XP_006713012.1
13, 14
2200
XM_006712950.2
XP_006713013.1
13, 14
2299
XM_006712951.2
XP_006713014.1
13, 14
2296
XM_006712952.2
XP_006713015.1
13, 14
2293
XM_006712953.1
XP_006713016.1
13, 14
2264
XM_017005615.1
XP_016861104.1
13, 14
2341
XM_017005616.1
XP_016861105.1
13, 14
2335
XM_017005617.1
XP_016861106.1
13, 14
2299
XM_017005618.1
XP_016861107.1
13, 14
2344
XM_017005619.1
XP_016861108.1
13, 14
2341
XM_017005620.1
XP_016861109.1
13, 14
2335
XM_017005621.1
XP_016861110.1
13, 14
2194
XM_017005622.1
XP_016861111.1
13, 14
2305
XM_017005623.1
XP_016861112.1
13, 14
2188
XM_017005624.1
XP_016861113.1
13, 14
2188
XM_017005625.1
XP_016861114.1
12, 13
2041
XM_017005626.1
XP_016861115.1
12, 13
2054
XM_017005627.1
XP_016861116.1
12, 13
2002
XM_017005628.1
XP_016861117.1
12, 13
1996
XM_017005629.1
XP_016861118.1
12, 13
2111
XM_017005630.1
XP_016861119.1
12, 13
2047
XM_017005631.1
XP_016861120.1
12, 13
2041
XM_017005632.1
XP_016861121.1
12, 13
2041
XM_017005633.1
XP_016861122.1
12, 13
2035
XM_017005634.1
XP_016861123.1
12, 13
2002
XM_017005635.1
XP_016861124.1
6, 7
778
GenBank mRNA
AB020698.1
-
12, 13
1882
AK294756.1
-
13, 14
2236
AK300425.1
-
13, 14
2167
BC014934.1
-
8, 9
1321
BC065909.1
-
1, 2
120
BC082241.1
-
10, 11
1416
BC142660.1
-
13, 14
2219
BC142727.1
-
13, 14
2225
BC146752.1
-
12, 13
1882
9007186
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
ER-associated ubiquitin-dependent protein catabolic process
regulation of protein stability
response to endoplasmic reticulum stress
negative regulation of skeletal muscle tissue development
protein stabilization
protein K48-linked deubiquitination
positive regulation of cell cycle process
regulation of cellular response to hypoxia
negative regulation of proteasomal protein catabolic process
regulation of ERAD pathway
thiol-dependent ubiquitin-specific protease activity
protein binding
ubiquitin protein ligase binding
metal ion binding
Hsp90 protein binding
Lys48-specific deubiquitinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0