Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286552.1 | NP_001273481.1 | 10 | 1076 | ||
NM_008733.4 | NP_032759.3 | 11 | 1168 | |||
NM_198059.3 | NP_932307.2 | 11 | 1168 | |||
GenBank mRNA | AK138925.1 | - | 10 | 1039 | ||
AY177622.1 | - | 11 | 1165 | |||
AY177623.1 | - | 11 | 1165 | |||
BC145109.1 | - | 10 | 983 | |||
U76618.1 | - | 2526 |