Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cux1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291233.1 | NP_001278162.1 | 5 | 928 | ||
NM_001291234.1 | NP_001278163.1 | 5 | 639 | |||
NM_001291238.1 | NP_001278167.1 | 5 | 639 | |||
NM_001291239.1 | NP_001278168.1 | 5 | 928 | |||
NM_001291240.1 | NP_001278169.1 | 5 | 928 | |||
NM_009986.4 | NP_034116.3 | 5 | 639 | |||
NM_198602.3 | NP_941004.2 | 5 | 639 | |||
GenBank mRNA | AB041595.1 | - | 5 | 775 | ||
AK098096.1 | - | 5 | 402 | |||
AK133543.1 | - | 5 | 402 | |||
AK144932.1 | - | 5 | 1299 | |||
AK220173.1 | - | 5 | 395 | |||
AY037807.1 | - | 5 | 369 | |||
BC014289.1 | - | 5 | 408 | |||
BC054370.1 | - | 5 | 362 | |||
BC079570.1 | - | 6 | 826 | |||
BC090847.1 | - | 5 | 411 | 30547458 | ||
CA317682.1 | - | 5 | 594 | |||
U68542.1 | - | 6 | 413 |