Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320546.1 | NP_001307475.1 | 2 | 1105 | ||
NM_001320547.1 | NP_001307476.1 | 2 | 476 | |||
NM_018031.4 | NP_060501.3 | 2 | 891 | |||
GenBank mRNA | AF099100.1 | - | 2 | 560 | ||
AK001080.1 | - | 2 | 560 | |||
AK225653.1 | - | 2 | 554 | |||
AK295145.1 | - | 2 | 891 | |||
AK296387.1 | - | 2 | 176 | |||
AK304117.1 | - | 2 | 476 | |||
AK309913.1 | - | 2 | 771 | |||
BC002826.2 | - | 460 | ||||
BC101707.1 | - | 2 | 550 | 8069213 | ||
BC113467.1 | - | 2 | 550 | 8322519 | ||
BC143486.1 | - | 2 | 550 | |||
DC300417.1 | - | 2 | 477 |