Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103171.1 | NP_001096641.1 | 22, 23 | 5766 | ||
NM_010862.2 | NP_034992.2 | 22, 23 | 5766 | |||
NM_182698.2 | NP_874357.2 | 21, 22 | 2004 | |||
GenBank mRNA | AF053130.1 | - | 20, 21 | 1972 | ||
AF144095.1 | - | 22, 23 | 5766 | |||
AY331132.1 | - | 22, 23 | 5766 | |||
AY331133.1 | - | 21, 22 | 2004 |