Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039071.2 | NP_001034160.1 | 9 | 1431 | ||
NM_001039072.2 | NP_001034161.1 | 11 | 1485 | |||
NM_001039073.2 | NP_001034162.1 | 11 | 1314 | |||
NM_001039074.2 | NP_001034163.1 | 11 | 1531 | |||
NM_011918.4 | NP_036048.3 | 10 | 1617 | |||
GenBank mRNA | AF114378.2 | - | 10 | 1583 | ||
AF228057.1 | - | 9 | 1510 | |||
AF228058.1 | - | 8 | 1324 | |||
AK084461.1 | - | 10 | 1600 | |||
AK142292.1 | - | 10 | 1569 | |||
AK172980.1 | - | 11 | 1531 | |||
AY206012.1 | - | 9 | 1397 | |||
AY206013.1 | - | 11 | 1449 | |||
AY206015.1 | - | 10 | 1230 | |||
BC138793.1 | - | 9 | 1551 | 8861815 | ||
BC145420.1 | - | 8 | 1365 | 9053978 |