Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009814.1 | NP_001009814.1 | 19, 20 | 2355 | ||
NM_015058.1 | NP_055873.1 | 19, 20 | 2355 | |||
XM_006719791.3 | XP_006719854.1 | 11, 12 | 1311 | |||
XM_011535007.2 | XP_011533309.1 | 19, 20 | 2381 | |||
XM_017020469.1 | XP_016875958.1 | 21, 22 | 2414 | |||
XM_017020470.1 | XP_016875959.1 | 19, 20 | 2381 | |||
XM_017020471.1 | XP_016875960.1 | 19, 20 | 2381 | |||
XM_017020472.1 | XP_016875961.1 | 3, 4 | 343 | |||
XM_017020474.1 | XP_016875963.1 | 19, 20 | 2381 | |||
GenBank mRNA | AB011136.1 | - | 8, 9 | 895 | ||
AK095155.1 | - | 19, 20 | 2229 | |||
AL834181.1 | - | 12, 13 | 1404 | |||
BC053674.1 | - | 19, 20 | 2355 | 6171968 | ||
BC131802.1 | - | 20, 21 | 2188 |