Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015060.2 | NP_055875.1 | 11 | 1589 | ||
XM_005249668.2 | XP_005249725.1 | 11 | 1526 | |||
XM_005249669.2 | XP_005249726.1 | 11 | 1526 | |||
XM_005249670.2 | XP_005249727.1 | 11 | 1232 | |||
XM_011515227.2 | XP_011513529.1 | 11 | 1526 | |||
XM_017011891.1 | XP_016867380.1 | 11 | 1232 | |||
XM_017011892.1 | XP_016867381.1 | 11 | 1242 | |||
GenBank mRNA | AK090619.1 | - | 11 | 1451 | ||
CN312870.1 | - | 1 | 101 | |||
D87682.1 | - | 9 | 1098 |