Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRUNE2 SIRNA Assays ›
Gene Symbol:
PRUNE2
Gene Name:
prune homolog 2
Gene Aliases:
BMCC1, BNIPXL, C9orf65, KIAA0367
Chromosome Location:
Chr.9: 76611376 - 76906220 on Build GRCh38
UniGene ID:
Hs.262857
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
261164
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRUNE2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308047.1
NP_001294976.1
18
10112
NM_001308048.1
NP_001294977.1
18
10115
NM_001308049.1
NP_001294978.1
12
2277
NM_001308050.1
NP_001294979.1
11
2132
NM_001308051.1
NP_001294980.1
9
2048
NM_015225.2
NP_056040.2
19
10190
NR_131751.1
16
9988
XM_005251745.1
XP_005251802.1
20
10335
XM_005251746.1
XP_005251803.1
20
10332
XM_005251748.1
XP_005251805.1
20
10329
XM_005251750.1
XP_005251807.1
19
10323
XM_005251751.1
XP_005251808.1
19
10284
XM_005251754.1
XP_005251811.1
17
10239
XM_006716982.1
XP_006717045.1
21
10371
XM_006716983.1
XP_006717046.1
21
10368
XM_006716985.1
XP_006717048.1
20
10362
XM_006716986.1
XP_006717049.1
20
10359
XM_011518323.1
XP_011516625.1
19
10326
XM_011518327.1
XP_011516629.1
19
10296
XM_017014345.1
XP_016869834.1
20
10332
XM_017014346.1
XP_016869835.1
19
10320
XM_017014347.1
XP_016869836.1
19
10293
XM_017014348.1
XP_016869837.1
18
10287
XM_017014349.1
XP_016869838.1
18
10284
XM_017014350.1
XP_016869839.1
19
10281
XM_017014351.1
XP_016869840.1
18
10275
XM_017014352.1
XP_016869841.1
18
10272
XM_017014354.1
XP_016869843.1
12
1960
XM_017014355.1
XP_016869844.1
11
1924
XM_017014356.1
XP_016869845.1
11
1921
XM_017014357.1
XP_016869846.1
10
1885
XM_017014358.1
XP_016869847.1
11
3778
XM_017014359.1
XP_016869848.1
10
1873
GenBank mRNA
AB002365.2
-
12
3262
AB050197.1
-
13
9296
BC150641.1
-
13
9208
9057011
FJ808772.1
-
18
10051
FJ808773.1
-
16
9927
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
pyrophosphatase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0