Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015354.2 | NP_056169.1 | 18, 19 | 1914 | ||
GenBank mRNA | BC040352.1 | - | 5, 6 | 521 | ||
BX537774.1 | - | 15, 16 | 1558 | |||
D79991.1 | - | 18, 19 | 1876 |