Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002814.2 | NP_001002814.2 | 6 | 4761 | ||
NM_025151.4 | NP_079427.4 | 5 | 2859 | |||
GenBank mRNA | AK026177.1 | - | 2 | 1100 | ||
AK092296.1 | - | 4 | 2167 | |||
AK122583.1 | - | 3 | 2900 | |||
AK124422.1 | - | 1 | 1659 | |||
AK226111.1 | - | 6 | 4451 | |||
AL833169.1 | - | 3 | 2171 | |||
BC077720.1 | - | 5 | 2829 | 6537110 |