Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027978.2 | NP_082254.2 | 3 | 480 | ||
GenBank mRNA | AK009092.1 | - | 3 | 482 | ||
AK200988.1 | - | 1 | 27 | |||
BC080773.1 | - | 3 | 475 | 30139077 |