Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
Tnnt3
Gene Name:
troponin T type 3 (skeletal, fast)
Gene Aliases:
Tnt
Chromosome Location:
Chr.1: 215666628 - 215683628 on Build Rnor_6.0
UniGene ID:
Rn.15488
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
261428
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnt3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270664.1
NP_001257593.1
12
639
NM_001270665.1
NP_001257594.1
14
672
NM_001270666.1
NP_001257595.1
14
672
NM_001270668.1
NP_001257597.1
12
639
NM_001270670.1
NP_001257599.1
13
654
NM_001270671.1
NP_001257600.1
13
654
NM_001270672.1
NP_001257601.1
12
645
NM_001270673.1
NP_001257602.1
13
657
NM_001270675.1
NP_001257604.1
12
645
NM_001270676.1
NP_001257605.1
13
657
NM_001270677.1
NP_001257606.1
11
627
NM_001270678.1
NP_001257607.1
13
660
NM_031532.2
NP_113720.1
13
660
XM_006230681.2
XP_006230743.1
12
602
XM_006230683.2
XP_006230745.1
11
590
XM_006230685.2
XP_006230747.1
10
572
XM_006230686.2
XP_006230748.1
10
572
XM_008760080.1
XP_008758302.1
15
706
XM_008760081.1
XP_008758303.1
15
706
XM_008760082.1
XP_008758304.1
14
656
XM_008760083.1
XP_008758305.1
14
653
XM_008760084.1
XP_008758306.1
13
679
XM_008760085.1
XP_008758307.1
13
644
XM_008760086.1
XP_008758308.1
12
626
GenBank mRNA
DQ062204.1
-
13
556
DQ273678.1
-
13
556
FQ214650.1
-
13
620
FQ214722.1
-
12
607
FQ214724.1
-
13
621
FQ214766.1
-
12
601
FQ214833.1
-
13
622
FQ215030.1
-
12
601
FQ215059.1
-
12
632
FQ215106.1
-
12
601
FQ215252.1
-
13
619
FQ215316.1
-
12
621
FQ215319.1
-
12
601
FQ215327.1
-
14
634
FQ215374.1
-
13
647
FQ215598.1
-
14
630
FQ215602.1
-
12
634
FQ215694.1
-
12
639
FQ215723.1
-
14
673
FQ215881.1
-
14
634
FQ215885.1
-
14
622
FQ215897.1
-
13
647
FQ215955.1
-
12
601
FQ216024.1
-
9
601
FQ216051.1
-
12
601
FQ216059.1
-
14
629
FQ216097.1
-
13
615
FQ216131.1
-
12
632
FQ216208.1
-
13
619
FQ216324.1
-
12
601
FQ216367.1
-
12
601
FQ216467.1
-
12
601
FQ216480.1
-
12
607
FQ216561.1
-
9
601
FQ216694.1
-
13
616
FQ216711.1
-
12
638
FQ216750.1
-
9
492
FQ216784.1
-
12
607
FQ216866.1
-
14
634
FQ216925.1
-
4
363
FQ217147.1
-
12
600
FQ217587.1
-
2
110
FQ217846.1
-
14
666
FQ217900.1
-
13
647
FQ223840.1
-
12
601
FQ223999.1
-
12
601
FQ224006.1
-
12
603
FQ224794.1
-
12
601
FQ224836.1
-
12
601
FQ224839.1
-
13
619
FQ224892.1
-
13
622
FQ224966.1
-
13
605
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
muscle contraction
regulation of muscle contraction
regulation of striated muscle contraction
metabolic process
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
troponin T binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0