Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018072.5 | NP_060542.4 | 30, 31 | 4468 | ||
GenBank mRNA | AK098212.1 | - | 5, 6 | 739 | ||
AK125663.1 | - | 12, 13 | 1685 | |||
AK304129.1 | - | 10, 11 | 1545 | |||
AK316413.1 | - | 10, 11 | 1439 | |||
BC035302.1 | - | 1, 2 | 43 | |||
BC065205.1 | - | 8, 9 | 1204 | |||
BC130000.1 | - | 15, 16 | 2392 | |||
BC150614.1 | - | 29, 30 | 4338 | 9056984 |