Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252570.1 | NP_001239499.1 | 6, 7 | 520 | ||
NM_001281829.1 | NP_001268758.1 | 7, 8 | 546 | |||
NM_172267.4 | NP_758471.1 | 7, 8 | 899 | |||
GenBank mRNA | AA620138.1 | - | 3, 4 | 141 | ||
AK005293.1 | - | 7, 8 | 900 | |||
AK138607.1 | - | 7, 8 | 574 | |||
BC039982.1 | - | 7, 8 | 541 | |||
BC132271.1 | - | 7, 8 | 510 | 40130548 | ||
BC132273.1 | - | 7, 8 | 510 | 40130550 | ||
BQ829083.1 | - | 6, 7 | 516 | |||
CX235839.1 | - | 5, 6 | 366 |