Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002257.2 | NP_001002257.1 | 6 | 4676 | ||
NM_001304445.1 | NP_001291374.1 | 8 | 5081 | |||
NM_001304446.1 | NP_001291375.1 | 6 | 4685 | |||
NM_182551.4 | NP_872357.2 | 7 | 4868 | |||
XM_005264244.1 | XP_005264301.1 | 7 | 4830 | |||
XM_011532744.2 | XP_011531046.1 | 7 | 4828 | |||
GenBank mRNA | AK023269.1 | - | 1 | 2412 | ||
BC146817.1 | - | 6 | 4676 | 8860419 | ||
BX537607.1 | - | 1 | 1287 | |||
BX647953.1 | - | 6 | 4644 |