Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198746.1 | NP_942041.1 | 4 | 499 | ||
XM_008763341.1 | XP_008761563.1 | 4 | 655 | |||
XM_008763342.1 | XP_008761564.1 | 4 | 475 | |||
XM_008763343.1 | XP_008761565.1 | 4 | 385 | |||
XM_008763344.1 | XP_008761566.1 | 686 | ||||
GenBank mRNA | U56863.1 | - | 4 | 499 |