Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26174
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077351.1
NP_001070819.1
3
303
NM_001077352.1
NP_001070820.1
3
303
NM_018107.4
NP_060577.3
3
303
XM_011536890.2
XP_011535192.1
4
386
XM_011536892.2
XP_011535194.1
4
385
XM_011536893.2
XP_011535195.1
4
386
XM_011536894.2
XP_011535196.1
4
386
XM_011536895.2
XP_011535197.1
4
410
XM_011536896.2
XP_011535198.1
4
386
XM_011536897.2
XP_011535199.1
3
718
XM_011536900.1
XP_011535202.1
3
300
XM_017021398.1
XP_016876887.1
4
388
XM_017021399.1
XP_016876888.1
4
389
XM_017021400.1
XP_016876889.1
3
253
XM_017021401.1
XP_016876890.1
3
300
XM_017021402.1
XP_016876891.1
3
211
XM_017021403.1
XP_016876892.1
3
277
XM_017021404.1
XP_016876893.1
3
227
XM_017021405.1
XP_016876894.1
3
300
XM_017021406.1
XP_016876895.1
3
718
XM_017021407.1
XP_016876896.1
3
227
XM_017021408.1
XP_016876897.1
3
265
XM_017021409.1
XP_016876898.1
3
718
XM_017021410.1
XP_016876899.1
3
287
XM_017021411.1
XP_016876900.1
3
718
XM_017021412.1
XP_016876901.1
4
386
XM_017021413.1
XP_016876902.1
2
114
GenBank mRNA
AF087905.1
-
3
254
AF275678.1
-
3
257
AK001344.1
-
3
253
AK125593.1
-
3
265
AK225792.1
-
3
265
AK295448.1
-
3
265
AK308055.1
-
3
266
AL834198.1
-
3
264
BC002566.2
-
3
281
3138909
BC024208.2
-
3
240
2960467
BX161440.1
-
3
236
CK000110.1
-
3
277
CR457257.1
-
2
104
DC363343.1
-
3
298
KU178687.1
-
2
104
KU178688.1
-
2
104
KU178689.1
-
2
104
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
nucleotide binding
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0