Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017477.1 | NP_001017477.1 | 6 | 896 | ||
XM_006246783.2 | XP_006246845.1 | 7 | 846 | |||
XM_006246785.1 | XP_006246847.1 | 7 | 846 | |||
XM_008767855.1 | XP_008766077.1 | 7 | 846 | |||
XM_008767856.1 | XP_008766078.1 | 7 | 846 | |||
GenBank mRNA | BC083699.1 | - | 6 | 896 | 7190067 | |
FQ233878.1 | - | 7 | 730 |