Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cxcl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017496.1 | NP_001017496.1 | 4 | 980 | ||
XM_006250710.2 | XP_006250772.1 | 4 | 1021 | |||
GenBank mRNA | BC088260.1 | - | 4 | 980 | 7375992 | |
FQ234803.1 | - | 2 | 770 |