Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018012.3 | NP_060482.2 | 15 | 6921 | ||
GenBank mRNA | AK001019.1 | - | 4 | 1860 | ||
AK125187.1 | - | 8 | 2376 | |||
AY923834.1 | - | 15 | 6922 |