Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289009.1 | NP_001275938.1 | 3 | 1382 | ||
NM_018042.4 | NP_060512.3 | 3 | 1447 | |||
XM_005257995.4 | XP_005258052.1 | 4 | 1338 | |||
XM_011524967.2 | XP_011523269.1 | 5 | 1460 | |||
XM_017024809.1 | XP_016880298.1 | 3 | 1271 | |||
XM_017024810.1 | XP_016880299.1 | 4 | 1928 | |||
XM_017024811.1 | XP_016880300.1 | 5 | 1460 | |||
GenBank mRNA | AK001122.1 | - | 3 | 1423 | ||
AK291826.1 | - | 3 | 1360 | |||
BC035605.1 | - | 3 | 1447 | 5163323 | ||
BX647081.1 | - | 6 | 1570 |