Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028735.3 | NP_083011.2 | 25 | 3438 | ||
GenBank mRNA | AK015017.1 | - | 25 | 3398 | ||
AK029952.1 | - | 25 | 3442 | |||
AK133304.1 | - | 25 | 3410 | |||
BC133699.1 | - | 25 | 3396 | 40098338 | ||
DQ011665.1 | - | 25 | 3442 |