Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028735.3 | NP_083011.2 | 17, 18 | 2445 | ||
GenBank mRNA | AK015017.1 | - | 17, 18 | 2405 | ||
AK029952.1 | - | 17, 18 | 2449 | |||
AK133304.1 | - | 17, 18 | 2417 | |||
AK163163.1 | - | 17, 18 | 2167 | |||
BC133699.1 | - | 17, 18 | 2403 | 40098338 | ||
DQ011665.1 | - | 17, 18 | 2449 |