Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc35d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029529.3 | NP_083805.1 | 2 | 1748 | ||
GenBank mRNA | AK018094.1 | - | 1 | 530 | ||
AK042761.1 | - | 2 | 1500 | |||
AK044973.1 | - | 2 | 1485 | |||
AK047610.1 | - | 2 | 1749 | |||
BC139194.1 | - | 2 | 1578 | 8862216 |