Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RADIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018059.4 | NP_060529.4 | 10, 11 | 2473 | ||
GenBank mRNA | AB058752.1 | - | 10, 11 | 2417 | ||
AK000997.1 | - | 6, 7 | 911 | |||
AK001186.1 | - | 7, 8 | 1456 | |||
AK294745.1 | - | 9, 10 | 2272 | |||
AK296325.1 | - | 9, 10 | 1994 | |||
AK302549.1 | - | 2465 | ||||
AL136731.1 | - | 5, 6 | 704 | |||
BC004919.2 | - | 1 | 324 | |||
BC117317.1 | - | 9, 10 | 2359 | 40125868 | ||
BC126311.1 | - | 9, 10 | 2359 | 8992027 | ||
BC143526.1 | - | 9, 10 | 2359 | 9052035 | ||
EF560723.1 | - | 9, 10 | 2382 |