Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029749.2 | NP_084025.2 | 13 | 1700 | ||
GenBank mRNA | AY336567.1 | - | 12 | 1573 | ||
BC137852.1 | - | 12 | 1593 | 8860871 | ||
CF532651.1 | - | 4 | 599 |