Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018105.2 | NP_060575.1 | 3 | 688 | ||
NM_199003.1 | NP_945354.1 | 2 | 492 | |||
GenBank mRNA | AK001339.1 | - | 3 | 682 | ||
AK223231.1 | - | 3 | 667 | |||
AL832077.1 | - | 2 | 466 | |||
BC021721.2 | - | 3 | 662 | 4795229 | ||
BG497522.1 | - | 3 | 689 | |||
CD639319.1 | - | 2 | 474 | |||
CR457256.1 | - | 3 | 451 |