Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
262782
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
2
247
NM_001018017.2
NP_001018017.1
2
220
NM_001044390.2
NP_001037855.1
2
220
NM_001044391.2
NP_001037856.1
2
220
NM_001044392.2
NP_001037857.1
2
247
NM_001044393.2
NP_001037858.1
2
220
NM_001204285.1
NP_001191214.1
2
220
NM_001204286.1
NP_001191215.1
2
247
NM_001204287.1
NP_001191216.1
2
247
NM_001204288.1
NP_001191217.1
2
247
NM_001204289.1
NP_001191218.1
2
247
NM_001204290.1
NP_001191219.1
2
184
NM_001204291.1
NP_001191220.1
2
247
NM_001204292.1
NP_001191221.1
2
247
NM_001204293.1
NP_001191222.1
2
220
NM_001204294.1
NP_001191223.1
2
220
NM_001204295.1
NP_001191224.1
2
247
NM_001204296.1
NP_001191225.1
2
247
NM_001204297.1
NP_001191226.1
2
247
NM_002456.5
NP_002447.4
2
220
GenBank mRNA
AF125525.1
-
2
139
AF348143.1
-
2
172
AK301575.1
-
2
214
AY327582.1
-
2
172
AY327583.1
-
2
172
AY327584.1
-
2
199
AY327585.1
-
2
172
AY327586.1
-
2
199
AY327587.1
-
2
225
AY327588.1
-
2
198
AY327589.1
-
2
172
AY327590.1
-
2
172
AY327591.1
-
2
172
AY327592.1
-
2
172
AY327595.1
-
2
172
AY327596.1
-
2
198
AY327597.1
-
2
172
AY327598.1
-
2
172
AY327599.1
-
2
199
AY327600.1
-
2
172
AY466157.1
-
2
148
BC120974.2
-
2
197
40115473
BC120975.2
-
2
224
40115475
EF583642.1
-
2
225
EF583643.1
-
2
324
EF583644.1
-
2
225
EF583645.1
-
2
324
EF583647.1
-
2
225
EF583648.1
-
2
225
EF583649.1
-
2
225
EF583650.1
-
2
162
EF583651.1
-
2
162
EF583652.1
-
2
225
EF583653.1
-
2
225
EF583654.1
-
2
162
EF583655.1
-
2
225
EF583656.1
-
2, 3
225
EF670706.1
-
2
189
EF670707.1
-
2
189
EF670708.1
-
2
225
EF670709.1
-
2
225
EF670710.1
-
2
198
EF670711.1
-
2
198
EF670712.1
-
2
198
EF670713.1
-
2
225
EF670714.1
-
2
198
EF670715.1
-
2
189
EF670716.1
-
2
225
EF670717.1
-
2
225
EF670718.1
-
2
225
EF670719.1
-
2
225
EF670720.1
-
2
162
EF670721.1
-
2
225
EF670722.1
-
2
162
EF670723.1
-
2
198
FJ226028.1
-
1
700
FJ226029.1
-
1
700
FJ226030.1
-
2
670
FJ226031.1
-
1
700
FJ226032.1
-
1
700
FJ226033.1
-
1
700
FJ226035.1
-
2
225
FJ226036.1
-
1
700
FJ226037.1
-
2
324
FJ226038.1
-
1
699
FJ226039.1
-
2
324
FJ226040.1
-
2
225
FJ226041.1
-
2
225
FJ226042.1
-
2
225
FJ226043.1
-
2
198
FJ226044.1
-
2
324
FJ226045.1
-
2
297
FJ226046.1
-
2
297
FJ226047.1
-
2
198
FJ226049.1
-
2
225
FJ226050.1
-
2
198
FJ226052.1
-
2
198
FJ226053.1
-
2
225
FJ226054.1
-
2
324
FJ226055.1
-
2
225
J05581.1
-
2
220
J05582.1
-
221
M31823.1
-
2
241
M32738.1
-
2
225
M32739.1
-
2
198
S81736.1
-
2
173
S81781.1
-
2
184
U60259.1
-
2
173
U60260.1
-
2
173
U60261.1
-
2
173
X52228.1
-
2
205
X52229.1
-
2
205
X80761.1
-
2
148
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline