Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sstr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009216.3 | NP_033242.1 | 2 | 1481 | ||
GenBank mRNA | AK046464.1 | - | 2 | 1481 |
Set Membership: |
![]() |