Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291128.1 | NP_001278057.1 | 3 | 1073 | ||
NM_001291129.1 | NP_001278058.1 | 3 | 1061 | |||
NM_001291130.1 | NP_001278059.1 | 3 | 1012 | |||
NM_010923.3 | NP_035053.1 | 3 | 1066 | |||
NM_180960.3 | NP_851291.1 | 2 | 985 | |||
GenBank mRNA | AB004048.1 | - | 3 | 1273 | ||
AK003004.1 | - | 3 | 1030 | |||
AK160626.1 | - | 3 | 1034 | |||
BC036984.1 | - | 3 | 1024 | 4981517 | ||
BU705167.1 | - | 1 | 327 | |||
CJ302750.1 | - | 1 | 240 | |||
GQ184462.1 | - | 3 | 968 | |||
X83568.1 | - | 3 | 834 | |||
X83569.1 | - | 2 | 941 | |||
X83570.1 | - | 3 | 1022 |