Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026352.3 | NP_080628.1 | 8 | 1017 | ||
GenBank mRNA | AK003402.1 | - | 8 | 974 | ||
AK012028.1 | - | 2 | 152 | |||
AK013919.1 | - | 8 | 1008 | |||
AK016221.1 | - | 8 | 1012 | |||
AK051597.1 | - | 8 | 997 | |||
AK142413.1 | - | 7 | 3441 | |||
AK150520.1 | - | 8 | 1034 | |||
AK151091.1 | - | 8 | 1034 | |||
BC011499.1 | - | 8 | 1014 | 3584183 | ||
BC019778.1 | - | 8 | 1017 | 4035785 |