Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASUN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018164.2 | NP_060634.2 | 2 | 663 | ||
XM_005253422.3 | XP_005253479.1 | 2 | 622 | |||
XM_011520750.2 | XP_011519052.1 | 2 | 196 | |||
XM_011520751.2 | XP_011519053.1 | 2 | 622 | |||
XM_011520752.2 | XP_011519054.1 | 2 | 622 | |||
XM_017019632.1 | XP_016875121.1 | 2 | 196 | |||
XM_017019633.1 | XP_016875122.1 | 2 | 663 | |||
XM_017019634.1 | XP_016875123.1 | 2 | 663 | |||
XM_017019635.1 | XP_016875124.1 | 2 | 196 | |||
XM_017019636.1 | XP_016875125.1 | 2 | 622 | |||
GenBank mRNA | AK001492.1 | - | 2 | 292 | ||
AK001499.1 | - | 2 | 328 | |||
AK225336.1 | - | 2 | 314 | |||
BC003081.2 | - | 2 | 352 | 2989678 | ||
BC008368.1 | - | 2 | 310 | 3355282 |