Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APITD1-CORT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270517.1 | NP_001257446.1 | 2 | 580 | ||
NM_198544.3 | NP_940946.1 | 2 | 580 | |||
NM_199006.2 | NP_950171.2 | 2 | 580 | |||
GenBank mRNA | AF516753.1 | - | 2 | 298 | ||
AF521016.1 | - | 2 | 580 | |||
HY054077.1 | - | 2 | 250 |
Gene Symbol: APITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199294.2 | NP_954988.1 | 2 | 580 | ||
NR_036462.1 | 2 | 768 | ||||
GenBank mRNA | BC029430.1 | - | 2 | 239 | 4051739 | |
BU902753.1 | - | 1 | 80 |