Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144856.1 | NP_001138328.1 | 3, 4 | 532 | ||
NM_001144857.1 | NP_001138329.1 | 2, 3 | 700 | |||
NM_018173.3 | NP_060643.2 | 3, 4 | 450 | |||
XM_005253704.4 | XP_005253761.1 | 3, 4 | 547 | |||
XM_006718985.3 | XP_006719048.1 | 3, 4 | 547 | |||
XM_011520967.2 | XP_011519269.1 | 3, 4 | 422 | |||
XM_017019555.1 | XP_016875044.1 | 3, 4 | 547 | |||
GenBank mRNA | AK001527.1 | - | 3, 4 | 451 | ||
AK023741.1 | - | 3, 4 | 532 | |||
AK095373.1 | - | 2, 3 | 700 | |||
BC104751.1 | - | 3, 4 | 386 | 40035012 | ||
BC105961.1 | - | 3, 4 | 386 | 40035014 | ||
BC109095.2 | - | 3, 4 | 386 | 40034695 |