Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM222B SIRNA Assays ›
Gene Symbol:
FAM222B
Gene Name:
family with sequence similarity 222 member B
Gene Aliases:
C17orf63
Chromosome Location:
Chr.17: 28755978 - 28855036 on Build GRCh38
UniGene ID:
Hs.564533
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26328
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM222B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077498.2
NP_001070966.1
3
1884
NM_001288631.1
NP_001275560.1
4
2055
NM_001288632.1
NP_001275561.1
5
1966
NM_001288633.1
NP_001275562.1
4
1960
NM_001288634.1
NP_001275563.1
3
1953
NM_001288635.1
NP_001275564.1
4
1932
NM_001288636.1
NP_001275565.1
6
1956
NM_001288637.1
NP_001275566.1
5
1922
NM_001288638.1
NP_001275567.1
5
1854
NM_001288639.1
NP_001275568.1
4
1806
NM_001288640.1
NP_001275569.1
3
1772
NM_018182.3
NP_060652.2
4
1918
XM_011524986.2
XP_011523288.1
3
1849
XM_011524987.1
XP_011523289.1
4
1684
XM_011524988.2
XP_011523290.1
3
1900
XM_017024831.1
XP_016880320.1
4
2008
XM_017024832.1
XP_016880321.1
3
2273
XM_017024833.1
XP_016880322.1
3
1813
XM_017024834.1
XP_016880323.1
3
2011
XM_017024835.1
XP_016880324.1
4
2210
GenBank mRNA
AK000147.1
-
1
1028
AK001562.1
-
4
1869
AK025980.1
-
1
887
AK057665.1
-
3
1868
AK095569.1
-
3
1953
AK299477.1
-
3
1718
BC021228.2
-
3
1768
4040385
BX647766.1
-
3
1703
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-sq56x:80/10.239.161.141:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline